Methyl-4-(bromomethyl)benzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 4-(bromomethyl)benzoate

072778-100G 072778-25G
102526-696EA 203.08 USD
102526-696 102833-548
Methyl-4-(bromomethyl)benzoate ≥95%
Methyl 4-(bromomethyl)benzoate

Hazard: Corrosive

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 130…135 °C (2 mmHg)
Melting Pt: 53…58 °C
MDL Number: MFCD00032453
CAS Number: 2417-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR