Imidazole-4-carboxylic acid ethyl ester ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl imidazole-4-carboxylate

072776-1G 072776-5G
102526-682EA 16.92 USD
102526-682 102526-684
Imidazole-4-carboxylic acid ethyl ester ≥95%
Imidazole-4-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
MDL Number: MFCD07363812
CAS Number: 23785-21-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR