2-Amino-5-nitro-6-picoline ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-methyl-3-nitropyridine, 2-Amino-5-nitro-6-methylpyridine, 2-Amino-6-methyl-5-nitropyridine, 6-Amino-3-nitro-2-picoline, 6-Methyl-5-nitro-2-pyridylamine

072773-1G 072773-5G 072773-10G
102526-666EA 27.69 USD
102526-666 102526-668 102526-670
2-Amino-5-nitro-6-picoline ≥95%
2-Amino-5-nitro-6-picoline

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14
MDL Number: MFCD00053582
CAS Number: 22280-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR