2-Amino-6-methyl-3-nitropyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Methyl-3-nitro-2-pyridylamine, 6-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-6-picoline, 2-Amino-3-nitro-6-picoline

072772-1G 072772-5G 072772-10G
102526-660EA 72.31 USD
102526-660 102526-662 102526-664
2-Amino-6-methyl-3-nitropyridine ≥95%
2-Amino-6-methyl-3-nitropyridine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD00047443
CAS Number: 21901-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR