2-Amino-4,6-dimethoxybenzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-amino-4,6-dimethoxybenzoic acid

072771-1G 072771-5G 072771-25G
102526-654EA 123.08 USD
102526-654 102526-656 102663-894
2-Amino-4,6-dimethoxybenzoic acid ≥95%
2-Amino-4,6-dimethoxybenzoic acid

Hazard: Irritant

Formula: C₉H₁₁NO₄
MW: 197.1900 g/mol
MDL Number: MFCD09031989
CAS Number: 21577-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR