Methyl-5-bromo-3-methylpicolinate ≥95%
Supplier: Matrix Scientific
Synonyms:
5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester, Methyl 5-bromo-3-methylpyridine-2-carboxylate, Methyl 5-bromo-3-methylpicolinate
072770-1G
072770-5G
072770-10G
102526-648EA
355.38
USD
102526-648
102526-650
102526-652
Methyl-5-bromo-3-methylpicolinate ≥95%
Methyl 5-bromo-3-methylpicolinate
Hazard: Irritant
Formula:
C₈H₈BrNO₃ MW: 230.0586 g/mol |
MDL Number:
MFCD07375126 CAS Number: 213771-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...