Ethyl-6-bromopicolinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-bromopyridine-2-carboxylate

072768-5G 072768-10G 072768-100G
102526-636EA 84.62 USD
102526-636 102526-638 102526-640
Ethyl-6-bromopicolinate ≥95%
Ethyl 6-bromopicolinate

Hazard: Irritant

Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
MDL Number: MFCD04116913
CAS Number: 21190-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR