2,3-Dichloro-6-nitrobenzonitrile ≥95%
Supplier: Matrix Scientific
072767-5G
072767-10G
072767-100G
102526-630EA
107.69
USD
102526-630
102526-632
102526-634
2,3-Dichloro-6-nitrobenzonitrile ≥95%
2,3-Dichloro-6-nitrobenzonitrile
Hazard: Irritant
Formula:
C₇H₂Cl₂N₂O₂ MW: 217.01 g/mol |
MDL Number:
MFCD00728596 CAS Number: 2112-22-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...