2-Iodopyridin-3-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-2-iodopyridine

072764-250MG 072764-1G 072764-5G
102526-614EA 73.85 USD
102526-614 102526-616 102526-618
2-Iodopyridin-3-amine ≥95%
2-Iodopyridin-3-amine

Hazard: Irritant

Formula: C₅H₅IN₂
MW: 220.0100 g/mol
MDL Number: MFCD07779506
CAS Number: 209286-97-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR