2-Fluoro-6-formylpyridine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Fluoropicolinaldehyde, 6-Fluoropyridine-2-carboxaldehyde
072763-1G
072763-5G
102526-610EA
135.38
USD
102526-610
102526-612
2-Fluoro-6-formylpyridine ≥95%
2-Fluoro-6-formylpyridine
Hazard: Irritant
Formula:
C₆H₄FNO MW: 125.1 g/mol |
MDL Number:
MFCD03095149 CAS Number: 208110-81-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...