5-Chloro-3-methylanthranilic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-chloro-3-methylbenzoic acid

072762-100G 072762-25G
102526-608EA 133.85 USD
102526-608 102551-802
5-Chloro-3-methylanthranilic acid ≥95%
5-Chloro-3-methylanthranilic acid

Hazard: Irritant

Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD02358895
CAS Number: 20776-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR