6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate ≥95%

Supplier: Matrix Scientific

072760-1G 072760-5G 072760-10G
102526-594EA 26.15 USD
102526-594 102526-596 102526-598
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate ≥95%
6,7-Dimethoxy-3,4-dihydroisoquinoline hydrochloride hydrate

Hazard: Irritant

Formula: C₁₁H₁₃NO₂.HCl.xH₂O
MW: 227.69 g/mol (anhy)
MDL Number: MFCD11099370
CAS Number: 20232-39-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR