2,6-Dibromoisonicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2,6-Dibromopyridine-4-carboxylic acid

072759-1G 072759-5G
102526-588EA 44.62 USD
102526-588 102526-590
2,6-Dibromoisonicotinic acid ≥95%
2,6-Dibromoisonicotinic acid

Hazard: Irritant

Formula: C₆H₃Br₂NO₂
MW: 280.9000 g/mol
MDL Number: MFCD09743987
CAS Number: 2016-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR