3-Methoxy-o-toluidine ≥95%

Supplier: Matrix Scientific

072758-5G 072758-25G 072758-100G
102526-584EA 52.31 USD
102526-584 102663-890 102663-892
3-Methoxy-o-toluidine ≥95%
3-Methoxy-o-toluidine

Hazard: Irritant

Formula: C₈H₁₁NO
Melting Pt: 27-28°C
MDL Number: MFCD06412568
CAS Number: 19500-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR