Ethyl-1H-indazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl Indazole-5-carboxylate, Ethyl 1H-indazole-5-carboxylate, Indazole-5-carboxylic acid ethyl ester

072757-1G 072757-5G 072757-10G
102526-576EA 152.31 USD
102526-576 102526-578 102526-580
Ethyl-1H-indazole-5-carboxylate ≥95%
Ethyl 1H-indazole-5-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.20 g/mol
MDL Number: MFCD03426220
CAS Number: 192944-51-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR