5-Amino-2-fluoro-3-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2-fluoro-3-picoline, 6-Fluoro-5-methylpyridin-3-amine

072755-1G 072755-5G
102526-564EA 95.38 USD
102526-564 102526-566
5-Amino-2-fluoro-3-methylpyridine ≥95%
5-Amino-2-fluoro-3-methylpyridine

Hazard: Irritant

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03095088
CAS Number: 186593-48-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR