5-Chloro-2-fluorophenol ≥95%

Supplier: Matrix Scientific

072754-25G 072754-100G
102663-886EA 110.77 USD
102663-886 102663-888
5-Chloro-2-fluorophenol ≥95%
5-Chloro-2-fluorophenol

Hazard: Irritant

Formula: C₆H₄ClFO
MW: 146.55 g/mol
MDL Number: MFCD04116357
CAS Number: 186589-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR