Ethyl-6-hydroxynicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 6-hydroxynicotinate

072753-1G 072753-5G 072753-10G
102526-552EA 284.62 USD
102526-552 102526-554 102526-556
Ethyl-6-hydroxynicotinate ≥95%
Ethyl-6-hydroxynicotinate

Hazard: Irritant

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
MDL Number: MFCD08460993
CAS Number: 18617-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR