4-Hydroxy-2-methylpyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Methylpyridin-4-ol

072752-5G 072752-25G
102526-548EA 156.92 USD
102526-548 102663-884
4-Hydroxy-2-methylpyridine ≥95%
4-Hydroxy-2-methylpyridine

Hazard: Irritant

Formula: C₆H₇NO
MW: 109.1259 g/mol
MDL Number: MFCD04114156
CAS Number: 18615-86-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR