3-Methyl-4-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

072750-1G 072750-5G
102526-536EA 223.08 USD
102526-536 102526-538
3-Methyl-4-nitrobenzaldehyde ≥95%
3-Methyl-4-nitrobenzaldehyde

Hazard: Irritant

Formula: C8H7NO3
MW: 165.1500 g/mol
MDL Number: MFCD08445621
CAS Number: 18515-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR