4-Fluoro-2-nitrobenzotrifluoride ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Fluoro-2-nitro-1-(trifluoromethyl)benzene

072749-1G 072749-5G
102526-530EA 710.77 USD
102526-530 102526-532
4-Fluoro-2-nitrobenzotrifluoride ≥95%
4-Fluoro-2-nitrobenzotrifluoride

Hazard: Irritant

Formula: C₇H₃F₄NO₂
CAS Number: 182289-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR