7-(Benzyloxy)-4-chloroquinoline ≥95%

Supplier: Matrix Scientific

072748-250MG 072748-1G 072748-5G
102526-524EA 355.38 USD
102526-524 102526-526 102526-528
7-(Benzyloxy)-4-chloroquinoline ≥95%
7-(Benzyloxy)-4-chloroquinoline

Hazard: Irritant

Formula: C₁₆H₁₂ClNO
MW: 269.73 g/mol
MDL Number: MFCD06659039
CAS Number: 178984-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR