4,5,6-Trichloropyrimidine ≥95%

Supplier: Matrix Scientific

072747-1G 072747-5G
102526-520EA 380 USD
102526-520 102526-522
4,5,6-Trichloropyrimidine ≥95%
4,5,6-Trichloropyrimidine

Hazard: Irritant

Formula: C₄HCl₃N₂
MW: 183.4231 g/mol
MDL Number: MFCD08458457
CAS Number: 1780-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR