tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

072746-1G 072746-5G 072746-10G
102526-512EA 80 USD
102526-512 102526-514 102526-516
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate ≥95%
tert-Butyl-2-(aminomethyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD01317793
CAS Number: 177911-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR