(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥95%

Supplier: Matrix Scientific

072745-5G 072745-25G
102526-508EA 30.77 USD
102526-508 102833-544
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone ≥95%
(S)-(+)-5-Hydroxymethyl-2-pyrrolidinone

Hazard: Irritant

Formula: C₅H₉NO₂
MW: 115.13 g/mol
Melting Pt: 78…82 °C
MDL Number: MFCD00077792
CAS Number: 17342-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR