3-Bromo-4-methoxyphenol ≥95%

Supplier: Matrix Scientific

072744-1G 072744-5G
102526-502EA 78.46 USD
102526-502 102526-504
3-Bromo-4-methoxyphenol ≥95%
3-Bromo-4-methoxyphenol

Hazard: Irritant

Formula: C7H7BrO2
MW: 203.0300 g/mol
MDL Number: MFCD11054143
CAS Number: 17332-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR