Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione ≥95%

Supplier: Matrix Scientific

072743-250MG 072743-1G
102526-498EA 1223.08 USD
102526-498 102526-500
Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione ≥95%
Pyrimido[4,5-d]pyridazine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C₆H₄N₄O₂
MDL Number: MFCD11111471
CAS Number: 17257-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR