4-Bromo-2-chlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-bromobenzaldehyde

072737-5G 072737-25G 072737-100G
102526-468EA 36.92 USD
102526-468 102833-540 102833-542
4-Bromo-2-chlorobenzaldehyde ≥95%
4-Bromo-2-chlorobenzaldehyde

Hazard: Irritant/Air Sensitive

Formula: C₇H₄BrClO
MW: 219.46 g/mol
MDL Number: MFCD08741413
CAS Number: 158435-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR