Methyl-2-(bromomethyl)-4-methoxybenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(bromomethyl)-4-methoxybenzoate

072736-1G 072736-5G
102526-462EA 238.46 USD
102526-462 102526-464
Methyl-2-(bromomethyl)-4-methoxybenzoate ≥95%
Methyl 2-(bromomethyl)-4-methoxybenzoate

Hazard: Irritant

Formula: C₁₀H₁₁BrO₃
MW: 259.1000 g/mol
MDL Number: MFCD04114316
CAS Number: 15365-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR