5-Hydroxynicotinonitrile ≥95%

Supplier: Matrix Scientific

072735-1G 072735-5G
102526-458EA 340 USD
102526-458 102526-460
5-Hydroxynicotinonitrile ≥95%
5-Hydroxynicotinonitrile

Hazard: Irritant

Formula: C6H4N2O
MW: 120.11 g/mol
MDL Number: MFCD11044307
CAS Number: 152803-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR