cis-Octahydropyrrolo[3,4-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: cis-Octahydro-1H-pyrrolo[3,4-b]pyridine

072734-1G 072734-5G 072734-10G
102526-452EA 366.15 USD
102526-452 102526-454 102526-456
cis-Octahydropyrrolo[3,4-b]pyridine ≥95%
cis-Octahydropyrrolo[3,4-b]pyridine

Hazard: Irritant

Formula: C₇H₁₄N₂
MW: 126.2 g/mol
MDL Number: MFCD04116298
CAS Number: 147459-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR