1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: Cyclopropyl 2-Fluorobenzyl ketone

072733-1G 072733-5G 072733-10G
102526-446EA 69.23 USD
102526-446 102526-448 102526-450
1-Cyclopropyl-2-(2-fluorophenyl)ethanone ≥95%
1-Cyclopropyl-2-(2-fluorophenyl)ethanone

Hazard: Irritant

Formula: C₁₁H₁₁FO
MW: 178.2000 g/mol
MDL Number: MFCD10687164
CAS Number: 150322-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR