(S)-2,3-Diaminopropanoic acid hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: H-Dap-OH.HCl, H-Dap-OH·HCl, L-3-Aminoalanine Hydrochloride, (S)-2,3-Diaminopropionic acid monohydrochloride

072732-1G 072732-5G 072732-10G
102526-440EA 67.69 USD
102526-440 102526-442 102526-444
(S)-2,3-Diaminopropanoic acid hydrochloride ≥95%
(S)-2,3-Diaminopropanoic acid hydrochloride

Hazard: Irritant

Formula: C₃H₈N₂O₂·HCl
MW: 140.6 g/mol
MDL Number: MFCD00065497
CAS Number: 1482-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR