3,5-Bis(benzyloxy)benzaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 3,5-Dibenzyloxybenzaldehyde

072730-1G 072730-5G 072730-10G
102526-428EA 52.31 USD
102526-428 102526-430 102526-432
3,5-Bis(benzyloxy)benzaldehyde ≥95%
3,5-Bis(benzyloxy)benzaldehyde

Hazard: Irritant

Formula: C₂₁H₁₈O₃
MW: 318.3658 g/mol
MDL Number: MFCD00075777
CAS Number: 14615-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR