Cycloheptanecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Cycloheptane carboxylic acid

072729-1G 072729-5G 072729-10G
102526-422EA 38.46 USD
102526-422 102526-424 102526-426
Cycloheptanecarboxylic acid ≥95%
Cycloheptanecarboxylic acid

Hazard: Irritant

Formula: C₈H₁₄O₂
MW: 142.2 g/mol
Boiling Pt: 135…138 °C (19 mmHg)
Density: 1.035
Flash Pt: >113 °C
MDL Number: MFCD00004152
CAS Number: 1460-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR