2-Amino-3,5-dimethylbenzoic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3,5-Dimethylanthranilic acid
072728-1G
072728-5G
072728-10G
102526-416EA
60
USD
102526-416
102526-418
102526-420
2-Amino-3,5-dimethylbenzoic acid ≥95%
2-Amino-3,5-dimethylbenzoic acid
Hazard: Irritant
Formula:
C₉H₁₁NO₂ MW: 165.19 g/mol |
MDL Number:
MFCD00017099 CAS Number: 14438-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...