2-(3-Aminophenyl)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Aminophenylacetic acid

072726-1G 072726-5G
102526-404EA 36.92 USD
102526-404 102526-406
2-(3-Aminophenyl)acetic acid ≥95%
2-(3-Aminophenyl)acetic acid

Hazard: Irritant/Light Sensitive/Refrigerate

Formula: C₈H₉NO₂
MW: 151.1626 g/mol
Melting Pt: 147…150 °C
MDL Number: MFCD00075058
CAS Number: 14338-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR