Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 1-((2'-cyano-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

072725-1G 072725-5G 072725-10G
102526-398EA 213.85 USD
102526-398 102526-400 102526-402
Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate ≥95%
Ethyl-1-((2'-cyano[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

Hazard: Irritant

Formula: C₂₆H₂₃N₃O₃
MW: 425.4800 g/mol
MDL Number: MFCD09030636
CAS Number: 139481-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR