Benzyl-4-formylpiperidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Benzyl 4-formylpiperidine-1-carboxylate, 4-Formylpiperidine, N-CBZ protected

072723-5G 072723-25G
102526-388EA 81.54 USD
102526-388 102526-390
Benzyl-4-formylpiperidine-1-carboxylate ≥95%
Benzyl-4-formylpiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MW: 247.2897 g/mol
MDL Number: MFCD01317806
CAS Number: 138163-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR