(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: [R]-1-BOC-3-Pyrrolidinemethanol, (R)-tert-Butyl 3-(hydroxymethyl)pyrrolidine-1-carboxylate

072722-1G 072722-5G
102526-382EA 235.38 USD
102526-382 102526-384
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate ≥95%
(R)-tert-Butyl-3-(hydroxymethyl)pyrrolidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD03094725
CAS Number: 138108-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR