(E)-3-(Dimethylamino)-2-methoxyacrylaldehyde ≥95%

Supplier: Matrix Scientific

072721-250MG 072721-1G 072721-5G
102526-376EA 498.46 USD
102526-376 102526-378 102526-380
(E)-3-(Dimethylamino)-2-methoxyacrylaldehyde ≥95%
(E)-3-(Dimethylamino)-2-methoxyacrylaldehyde

Hazard: Irritant

Formula: C₆H₁₁NO₂
CAS Number: 13616-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR