(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(+)-3-(Dimethylamino)pyrrolidine, (R)-N,N-Dimethylpyrrolidin-3-amine

072717-250MG 072717-1G 072717-5G
102526-354EA 355.38 USD
102526-354 102526-356 102526-358
(3R)-(+)-3-(Dimethylamino)pyrrolidine ≥95%
(3R)-(+)-3-(Dimethylamino)pyrrolidine

Hazard: Irritant

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD00191347
CAS Number: 132958-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR