[(5-Methyl-1,3,4-oxadiazol-2-yl)methyl]amine ≥95%

Supplier: Matrix Scientific

072709-250MG 072709-1G 072709-5G
102526-306EA 273.85 USD
102526-306 102526-308 102526-310
[(5-Methyl-1,3,4-oxadiazol-2-yl)methyl]amine ≥95%
[(5-Methyl-1,3,4-oxadiazol-2-yl)methyl]amine

Hazard: Irritant

Formula: C₄H₇N₃O
CAS Number: 125295-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR