2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde ≥95%

Supplier: Matrix Scientific

072707-1G 072707-5G 072707-10G
102526-294EA 427.69 USD
102526-294 102526-296 102526-298
2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde ≥95%
2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carbaldehyde

Hazard: Irritant

Formula: C₁₉H₁₄FNO
MW: 291.3190 g/mol
MDL Number: MFCD09032925
CAS Number: 121660-37-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR