3,4-Dimethoxyphenethylamine ≥95%
Supplier: Matrix Scientific
072706-10G
072706-100G
102526-290EA
55.38
USD
102526-290
102526-292
3,4-Dimethoxyphenethylamine ≥95%
3,4-Dimethoxyphenethylamine
Hazard: Irritant
Formula:
C₁₀H₁₅NO₂ |
MDL Number:
MFCD00008188 CAS Number: 120-20-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...