3,4-Dihydroisoquinolin-1(2H)-one ≥95%
Supplier: Matrix Scientific
072705-1G
072705-5G
102526-286EA
141.54
USD
102526-286
102526-288
3,4-Dihydroisoquinolin-1(2H)-one ≥95%
3,4-Dihydroisoquinolin-1(2H)-one
Hazard: Irritant
Formula:
C9H9NO Boiling Pt: 388 °C Density: 1.142 Flash Pt: 226 °C |
MDL Number:
MFCD00853963 CAS Number: 1196-38-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...