2,4-Dichloro-D-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: H-D-Phe(2,4-DiCl)-OH

072703-1G 072703-5G 072703-10G
102526-272EA 320 USD
102526-272 102526-274 102526-276
2,4-Dichloro-D-phenylalanine ≥95%
2,4-Dichloro-D-phenylalanine

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MW: 234.09 g/mol
MDL Number: MFCD01860873
CAS Number: 114872-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR