4-Fluoro-L-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH

072701-1G 072701-5G 072701-10G
102526-260EA 1710.77 USD
102526-260 102526-262 102526-264
4-Fluoro-L-phenylalanine ≥95%
4-Fluoro-L-phenylalanine

Hazard: Irritant

Formula: C₉H₁₀FNO₂
MW: 183.2 g/mol
MDL Number: MFCD00063064
CAS Number: 1132-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR