5,6,7,8-Tetrahydroquinoline ≥95%
Supplier: Matrix Scientific
072692-10G
072692-100G
102526-212EA
143.08
USD
102526-212
102526-214
5,6,7,8-Tetrahydroquinoline ≥95%
5,6,7,8-Tetrahydroquinoline
Hazard: Irritant
Formula:
C₉H₁₁N MW: 133.19 g/mol |
MDL Number:
MFCD00006734 CAS Number: 10500-57-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...