5-Fluoro-2-methoxy-4-pyridineboronic acid ≥95%
Supplier: Matrix Scientific
072691-1G
072691-5G
102526-208EA
149.23
USD
102526-208
102526-210
5-Fluoro-2-methoxy-4-pyridineboronic acid ≥95%
5-Fluoro-2-methoxy-4-pyridineboronic acid
Hazard: Irritant
Formula:
C₆H₇BFNO₃ |
MDL Number:
MFCD06657887 CAS Number: 1043869-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...